Cardboard Freezer Storage Components, GeneMate
Supplier: GeneMate
D49 B2C D81 D100 D50 B3C
490000-576EA 6.98 USD
490000-576 490000-573 490000-574 490014-764 490000-570 490006-592
Cardboard Freezer Storage Components, GeneMate
Boxes Cryoboxes
Cardboard freezer boxes are for upright freezer and chest freezer racks
Boxes are available with or without dividers included. Dividers are also available separately to allow customized storage configurations.