GeneMate Flipper® Junior Microtube Racks, Scientific Specialties
Supplier: Scientific Specialties
5300-29-22IS1
490003-994BX 151.69 USD
490003-994
GeneMate Flipper® Junior Microtube Racks, Scientific Specialties
Racks Tube Racks
This compact rack is small enough to pick up with one hand, but big enough to hold enough tubes for most experiments. One side of the rack has 32 wells for 0.5 mL tubes plus an additional 8 wells across for 1.5/2.0 mL tubes. The reverse/flip side has the opposite configuration of 32 wells for 1.5/2.0 mL tubes and 8 wells for 0.5 mL tubes.
  • Polypropylene
  • Labeled with large easy-to-read 1-8 by A-D matrix
  • Dimensions: 3 1/8 x 6 1/4
  • Side A: 32 x 0.5 mL and 8 x 1.5/2.0 mL
  • Side B: 32 x 1.5/2.0 mL and 8 x 0.5 mL