Flipper Freezer Storage Racks, 64-Place, Scientific Specialties
Supplier: GeneMate
490004-000 490004-020 490004-034 490004-040 490004-052
490004-000BX 150.68 USD
490004-000 490004-020 490004-034 490004-040 490004-052
Flipper Freezer Storage Racks, 64-Place, Scientific Specialties
Racks Cryogenic Vial Racks
The Flipper Freezer Storage Rack is designed to work with most cryovials, 0.5, 1.5, 2.0 mL, and screw cap microtubes
This rack is designed to replace the 5 1/4 x 5 1/4" cardboard freezer storage boxes on the market. This Flipper freezer rack holds up to 64 tubes in an 8 x 8 matrix. The hole spacing is greater than with our standard 80-place rack making it possible to store tubes in all 64 holes simultaneously. Produced from copolymer polypropylene, this rack will withstand limited autoclaving (up to 15 minutes) as well as the rigors of deep freezer storage (down to -90 °C). Both sides areMore Product Information