Biochrom Colourwave CO7000 Medical Colorimeter
Supplier: Avantor
490005-900EA 2337.05 USD
490005-900
Biochrom Colourwave CO7000 Medical Colorimeter
Colorimeters Multiparameter Colorimeters
The Biochrom WPA CO7000 is a portable colorimeter designed for use by doctors and medical technologists in small and medium sized clinics.
  • Open system accommodates both conventional wet reagents and kits
  • Requires only a small volume of reagent (min. 0.4 mL) in a cuvette or tube
  • A portable, battery-operated unit you can take anywhere, ideal for field clinics
  • Fully tropicalized and portable
  • Reads assays in the wavelength range 400 to 700 nm using many proprietary test kits
  • Rechargeable battery
  • Registered for IVD applications
Easy, inexpensive colorimetry for the small medical clinic. The Colourwave CO7000 Medical Colorimeter is designed for use by doctors and medical technologists in small- and medium-sized clinics. It will withstand high temperature and humidity. The CO7000 is an open system that accommodates both conventional wet reagents and kits. The CO7000 requires only a small volume of reagent (min. 0.4 mL) in a cuvette or tube.A portable, battery-operated unit you can take anywhere, and is ideal for field clinics.
Ordering information: Comes with 1 year warranty.