Dynalon Bio-bin® Waste Disposal Containers
Supplier: Dynalon
Bio-bin®
797303-6300 797303-0002 797303-0030 797303-0006 797303-6200
490007-232EA 133.53 USD
490007-232 490007-226 490015-310 490007-228 490007-230
Dynalon Bio-bin® Waste Disposal Containers
Boxes Waste Disposal Boxes
Ideal use in academic, clinical, government and industrial laboratories for a wide rang of non-sharps biomedical and laboratory waste that require special heat treatment such as autoclaving or incineration before disposal.
  • Space efficient, lightweight fold-flat design
  • Ergonomic design, built-in lid and handles
  • Stackable post-use
  • Temporary & permanent seals
  • Printed user instructions & user ID/date information on lid
  • Waterproof construction
  • Built-in super absorbent Bio-matt to soak up any excess liquid
  • Autoclavable
6 liter Bio-bin is ideal for use with Biological Safety Cabinets.
Certifications: UN 3291 approved for handling Regulated Bio-Medical Waste.
Delivery information: Supplied with 2" x 2" Biohazard Waste Labels.