GeneMate 96-Well PCR Storage Racks, Scientific Specialties
Supplier: Scientific Specialties
5230-29-22IS1 5230-26-22IS1 5230-21-22IS1 5230-24-22IS1
490003-968BX 163.23 USD
490003-968 490003-978 490003-976 490003-974
GeneMate 96-Well PCR Storage Racks, Scientific Specialties
Racks Tube Racks
The rack is formatted in the standard 8 x 12 microtiter format and indexed A through H vertically and 1 through 12 horizontally for convenient and error-free tube identification. The included natural lids have a non-slip edge to allow non-slip stacking of multiple racks.
  • Fits individual tubes, 8- or 12-strips, and plates
  • Ideal for 0.2 mL tubes or strips
  • Packaged five per pack