Tube Storage Boxes, 15 and 50 ml, GeneMate
Supplier: GeneMate
490004-200 490004-202
490004-200BX 82.13 USD
490004-200 490004-202
Tube Storage Boxes, 15 and 50 ml, GeneMate
Boxes Cryoboxes
These polypropylene tube storage boxes are ideal for freezer storage.
  • Clear lid for easy location of tubes
  • Removable base for use in ice bath
  • Dimensions: 5.38×5.38×5.00" with lid
  • Frosted writing surface on two opposite lid sides for easy labeling
  • Green box holds 36 tubes
  • Blue box holds 16 tubes
  • White numerical indexing
  • 2 Racks/Pack