GeneMate Screw Cap Microtubes and Caps, Scientific Specialties
Supplier: VWR International
490003-344 490003-358 490003-362 490003-364 490003-366 490003-368 490003-370 490003-372 490003-374 490003-408 490003-420 490003-430
490003-344BX 202.32 USD
490003-344 490003-358 490003-362 490003-364 490003-366 490003-368 490003-370 490003-372 490003-374 490003-408 490003-420 490003-430
GeneMate Screw Cap Microtubes and Caps, Scientific Specialties
Tubes Microtubes
These screw cap microtubes offer the ultimate in applications from sample storage in vapor phase liquid nitrogen to boiling applications.
Manufactured under the most rigid QC standards, these tubes are certified to be RNAse, DNAse, DNA, and Pyrogen free. The O-Ring is manufactured from autoclavable ethelyne-propylene that forms a perfect seal against the rim of each tube. A leak free seal, coupled with the naturally low binding quality of 100% pure virgin copolymer polypropylene, make these screw cap tubes perfect for almost every long term storage application.
Ordering information: More Product Information