GeneMate LightCycler® Semi-Skirted, Low-Profile, 96-Well PCR Plates, Scientific Specialties
Supplier: GeneMate
3455-00-520IS 3455-40-520IS
490003-884BX 96.22 USD
490003-884 490003-886
GeneMate LightCycler® Semi-Skirted, Low-Profile, 96-Well PCR Plates, Scientific Specialties
Microplates PCR Plates
Designed to fit instruments using the LightCycler plates.
  • Ultra-thin wall for optimum heat transfer and higher yield
  • High contrast black alphanumerics for rapid sample identification
  • Plates can be sealed with strip caps, sealing mats, or sealing film
  • Crystal-clear clarity
  • Easy to visualize content of each well
  • Certified free from DNase, RNase, and detectable DNA
  • Also available either pre-barcoded or custom-barcoded for convenient sample identification and tracking
Available in both clear or white.