GeneMate Straight-sided, Semi-skirted 96-well PCR Plates, Scientific Specialties
Supplier: GeneMate
490003-866 490003-878
490003-866BX 96.96 USD
490003-866 490003-878
GeneMate Straight-sided, Semi-skirted 96-well PCR Plates, Scientific Specialties
Microplates PCR Plates
Compatible with most commercially available thermal cyclers.
  • Ultra-thin walls for optimum heat transfer and higher yield
  • High contrast black alphanumerics for rapid sample identification
  • Plates can be sealed with strip caps, sealing mats, or sealing film
  • Crystal-clear clarity
  • Easy to visualize contents of each well
  • Certified free from DNase, RNase, and detectable DNA
  • Available pre-barcoded or custom-barcoded for convenient sample identification and tracking
  • Available in a full range of colors