Standard and Specialty Petri Dishes, Bioplast Manufacturing
Supplier: Bioplast Manufacturing
2901 2917 2902 2900 2916 2901 2914 2910 2904 2906 2905
490005-226CS 374.41 USD
490005-226 490016-604 490005-214 490012-444 490012-222 490005-218 490005-216 490005-232 490005-220 490016-606 490005-230
Standard and Specialty Petri Dishes, Bioplast Manufacturing
Culture Dishes Petri Dishes
Petri dishes are used for all automated filling and streaking equipment.
  • Disposable
  • Radiation Sterilized
  • Precision and Convenience
  • Stackable, slippable dish construction
  • Ventilation ribs minimize condensation
Molding process produces optimum product clarity and the most consistently flat dish on the market. Can be traced for quality assurance and sterility verification. Side arrows and Isomarks available. Comprehensive line of standard, shallow, deep, and media saver dishes.