GeneMate Non-Skirted, 96-Well PCR Plate, Scientific Specialties
Supplier: GeneMate
3400-00-520IS 3400-09-520IS 3401-00-230IS
490003-750BX 128.06 USD
490003-750 490003-762 490003-770
GeneMate Non-Skirted, 96-Well PCR Plate, Scientific Specialties
Microplates PCR Plates
These plates are compatible with most commercially available thermal cyclers.
  • Ultra-thin wall for optimum heat transfer and higher yield
  • High contrast black alphanumerics for rapid sample identification
  • Plates can be sealed with strip caps, sealing mats, or sealing film
  • Crystal-clear clarity
  • Easy to visualize content of each well
  • Certified free from DNase, RNase, and detectable DNA
  • Cuttable; use only the wells you need
  • Available in white
  • Also available in 24-48 well configurations