8- Strip Ultraflux PCR Tubes and Caps, Scientific Specialties
Supplier: GeneMate
490003-528 490003-530 490003-534 490003-538 490003-540 490003-562 490003-564 490003-566 490003-572 490003-574 490003-596 490003-598 490003-600 490003-602 490003-604 490003-606 490004-430
490003-528BX 242.81 USD
490003-528 490003-530 490003-534 490003-538 490003-540 490003-562 490003-564 490003-566 490003-572 490003-574 490003-596 490003-598 490003-600 490003-602 490003-604 490003-606 490004-430
8- Strip Ultraflux PCR Tubes and Caps, Scientific Specialties
Tubes PCR Tubes
Ultraflux PCR tubes are designed to precisely fit most thermal cyclers.
  • Convenient tube and cap combination packs
  • Secure evaporation-resistant seal between cap lid and tube
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Tubes are pre-numbered for easy well identification
  • Crystal-clear clarity and easy to visualize content of each well
  • Ideal for enhancing signal strength for qPCR assays
Certified free DNase, RNase, and detectable DNA.
Ordering information: GeneMate 96-Well PCR tube storage racks are available separately.
Packaging: More Product Information