Microflex® N29 Nitrile Exam Gloves
Supplier: Ansell
Microflex®
N295 N292 N291 N290 N294 N291 N290 N293
10769-362CS 225.32 USD
CA10769-368 CA10769-366 CA10769-364 CA10769-362 CA10769-360 CA10769-370 10769-362 10769-360
Microflex® N29 Nitrile Exam Gloves
Gloves
These nitrile gloves are manufactured using a soft and stretchy formulation that offers excellent durability as well as comfort.
  • Powder-free
  • Textured for enhanced gripping power
  • Great tactile sensitivity
A three mil thickness provides enhanced tactile sensitivity.