GeneMate Sieve GQA Low Melt Agarose

Supplier: GeneMate
490001-604 490001-606 490001-608
490001-604EA 896.41 USD
490001-604 490001-606 490001-608
GeneMate Sieve GQA Low Melt Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate Sieve GQA Agarose is a low melting temperature agarose with the highest resolving capacity for DNA fragments smaller than 1000 bp, especially PCR products ranging from 200 to 800 bp. This agarose is Genetic Quality Analyzed, GQA, and carries that certification. This ensures that in-gel applications can be performed in remelted agarose, avoiding difficult DNA extraction steps. GeneMate Sieve GQA Agarose is ideal for digestion by agarose enzymes, making it very easy to recover large DNA fragments suitable for cloning or enzymatic processing. It can also be used at high concentrations,More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Ash Content 0.3%
DNase/RNase activity None detected
Gelling Temperature 35°C (4.0%)
Gel Strength 1000 g/cm2 (4%)
Enzymatic Degradation by Agarose Passes test
In-Gel Enzymatic Processing Passes test
Moisture 5%
Sulphate 0.12%
Melting Point 65°C, melting temperature (4.0%)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR