GeneMate HiRes Agarose
Supplier: GeneMate
MS-8-125G MS-8-25G MS-8-500G
490001-614EA 910.57 USD
490001-614 490001-618 490001-616
GeneMate HiRes Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate Hi-Res Agarose is specifically formulated for molecular screening and improves the resolution of small DNA fragments and PCR products. By harvesting the seaweed at the appropriate time in the maturity cycle and careful modifications to the polymer during manufacturing, we can bring you a product specially designed for applications that require efficient separation of small DNA fragments and PCR products. PCR process is covered by patents owned by Hoffman-LaRoche, Inc.
  • High Resolution of PCR and DNA Fragments with Low Base Pairs
  • Improved Clarity of the Gel, Enhancing Visibility
  • Better Handling, Stronger Gel Structure and Higher Gel Strength
  • Reduced Risk of Gel Cracking, Even at Low Concentrations
  • DNA binding: Very Low