Microvette® CB 300, Sarstedt
Supplier: SARSTEDT INC
16.443.100 16.444.100 16.444.100 16.440.100 16.443.100
101093-990PK 84.03 USD
101093-990 101093-988 101093-992 77587-382 77587-378 77587-380
Microvette® CB 300, Sarstedt
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The Microvette® is available for non-human use only.
  • Optimum serum or plasma recovery after centrifugation
  • Two in one sample container and centifuge tube
  • Compatible with different analyzers as a primary tube for direct sampling
The capillary tube with push cap is supplied complete with a carrier tube that is suitable for transport and centrifugation. Before Centrifugation, the capillary tube is sealed with the cap attached to the push cap. The trumpet-shaped capillary tube ensures maximum serum or plasma recovery after centrifugation. For efficiency in the laboratory, the capillary tube accomodated in its carrier tube can be placed directly onto the analyzer.