Accucore™ XL C18 4 μm HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
74104-254630 74104-253030 74104-154630 74104-153030 74104-104630 74104-103030 74104-102130 74104-052130
10038-664EA 1257.48 USD
10038-664 10038-666 10038-668 10038-670 10038-672 10038-674 10038-676 10038-678 10038-680 10038-682 10038-684 10038-686
Accucore™ XL C18 4 μm HPLC Columns, Thermo Scientific
Chromatography Columns
Accucore™ XL 4 μm C18 HPLC Columns provide optimum retention of non-polar analytes via a predominantly hydrophobic interaction mechanism.
  • 4 μm solid core particles for all users
  • Same system, same method, better results
  • Robust, fast, and easy to use
  • Optimum retention of non-polar compounds
  • Hydrophobic interaction mechanism
  • Separates a broad range of analytes
Based on Core Enhanced Technology™ using 4 μm solid core particles, Accucore™ XL HPLC columns allow users of conventional HPLC methods to enjoy performance far beyond that of columns packed with 5,More Product Information