TracePLOT TG-Bond Msieve 5A GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26003-6070 26003-1630 26003-6100 26003-6040 26003-6010
10056-026EA 2304.93 USD
10056-026 10056-018 10056-020 10056-024 10056-022
TracePLOT TG-Bond Msieve 5A GC Columns, Thermo Scientific
Chromatography Columns
Designed for separation of Ar/O₂ and other permanent gases.