TraceGOLD TG-1701MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26090-1420 26090-2960 26090-1540 26090-2350 26090-1440 26090-2240 26090-2230 26090-2980 26090-2970 26090-1430 26090-3080 26090-0630 26090-3960 26090-3090 26090-1550 26090-1310
10056-412EA 1178.53 USD
10056-412 10056-410 10056-416 10056-414 10055-904 10056-442 10056-396 10055-888 10055-902 10055-874 10055-884 10055-886 10055-892 10055-880 10056-408 10055-890
TraceGOLD TG-1701MS GC Columns, Thermo Scientific
Chromatography Columns
Feature a mix of cyano and phenyl groups for increased polarity and a different elution order relative to less polar columns.