Anti-MUC5AC Mouse Monoclonal Antibody [clone: 45M1]

Supplier: Diagnostic BioSystems

PDM163 MOB357-05 MOB357
10064-978EA 326.87 USD
10064-978 99990-668 95024-690
Anti-MUC5AC Mouse Monoclonal Antibody [clone: 45M1]
Antibodies
This antibody reacts with a 1000 kDa protein and recognizes the peptide core of gastric mucin M1. Mucins are high molecular weight glycoproteins with 80% carbohydrates and 20% core protein. Gastric mucin M1 antigen is found in columnar mucus cells of surface gastric epithelium and in goblet cells of the fetal and precancerous colon but not in normal colon. Resurgence of gastric mucin during colonic carcinogenesis is suggestive of either re-expression of the peptide core of gastric mucin in the adult colon or changes in the glycosylation pattern of mucin, which expose the hidden M1 antigen.

Type:
Antigen: MUC5AC
Clonality: Monoclonal
Clone: 45M1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR