HyperSep™ Protein Precipitation Plate, Thermo Scientific
Supplier: Thermo Scientific
60304-201
10048-502EA 230.32 USD
10048-502
HyperSep™ Protein Precipitation Plate, Thermo Scientific
Microplates Solid Phase Extraction Plates (SPE)
HyperSep™ Protein Precipitation Plates provide a quick, effective approach for removal of proteins from biological compounds using the protein crash technique.
  • Dual frit design
  • Hydrophobic/oleophobic frits enable only precipitation of proteins
  • Pore size is optimized for ideal flow rate
  • Specially selected PP for low extractables
In combination with SPE and SLE, the Protein Precipitation Plate offers a comprehensive range of options for sample preparation of biological based compounds. The 96-well plates are ideal for use in automated, high throughput systems.