Tenbroeck Tissue Grinder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64798-01 64798-02 64798-07 64798-15 64798-40
102095-690PK 301.16 USD
102095-690 102095-692 102095-694 102095-696 102095-698
Tenbroeck Tissue Grinder, Electron Microscopy Sciences
Homogenizers Tissue Grinders
All glass Tenbroeck tissue grinders are precision made and feature interchangeable pestles and tubes. No need to keep components matched, also offer easier replacement of damaged parts.
  • Ideal for grinding liver, heart, and intestine tissue
  • Made from Wheaton 33 borosilicate glass
  • Hollow pestle can be pack with ice
  • Pour lip on mortar – Autoclavable
The clearance between pestle and tube is 0.0035 to 0.0065" (0.09 – 0.16 mm). While this grinder is designed for hand operation, it can be slow speed motor-driven by inserting a rubber stopper (side 00) and metal rod into pestle. This unit also features an expanded reservoir and pouring lip.