Histosol™ Xylene Substitute, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
23405-01
23405-04
102092-290EA
163.97
USD
102092-290
102092-292
Histosol™ Xylene Substitute, Electron Microscopy Sciences
Stains
Microscopy and staining reagents
Histosol™ is the original high flash point (114°F TCC) histological clearing agent.
It is intended to be used as a replacement for xylene where the hazards associated with aromatic hydrocarbon vapors are to be reduced. Museum-quality tissue slides can be prepared with Hitosol without change in protocol or procedure. Histosol is manufactured from petrochemical products and is miscible in all proportions with ethanol, isopropanol, and t-butanol. It is also miscible with all paraffin-based tissue embedding media and all permanent mounting materials.