Antibody Saver Tray, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70480-10 70481-10 70481-20
102096-926PK 78.53 USD
102096-926 102096-928 102096-930
Antibody Saver Tray, Electron Microscopy Sciences
Trays/Dishes/Pots Laboratory Trays/Dishes
Antibody saver trays minimize the use of expensive antibodies when incubating filters in the Western Blot process.
  • Trays are available for popular midi and mini and multiple lane filters
  • Each tray is molded of polypropylene and includes an attached lid to protect from spillage
  • Trays are stackable, reclosable and reusable
  • Single lane trays are linked together for easier handling but can be separated