Mounting Media
Supplier: Electron Microscopy Sciences
17985-150
17985-200
102092-128EA
128.25
USD
102092-128
102092-134
Mounting Media
Stains
Microscopy and staining reagents
EMS Shield Mount is an aqueous non-permanent, low viscosity mounting medium for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyl rhodamine and Redox. The fluorescence is retained during storage at 4 °C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes.