EMS-Mount, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
17985-05
17985-06
102092-112EA
119.29
USD
102092-112
102092-114
102092-116
102092-118
EMS-Mount, Electron Microscopy Sciences
Stains
Microscopy and staining reagents
EMS-Mount is a non-permanent, low viscosity an aqueous mounting medium designed for mounting of tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents. This mounting medium preserves Fast red, Aminoethylycarbazole (AEC), BCIP/NBT, BCIP/INT chromogens and is also compatible with counterstain like Hematoxylin and Nuclear fast red (NFR). It is also suitable for chromogens like DAB and DAB with nickel and cobalt.
- Ready to use mounting medium
- Use for the mounting of chromogens that cannot be dehydrated with organic solvents
- Not compatible with H and E staining