Vacule Tubing Vials and Stoppers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60304-04 60304-05 60304-10
102680-960PK 390.21 USD
102680-960 102680-962 102680-964
Vacule Tubing Vials and Stoppers, Electron Microscopy Sciences
Vials
Heavy wall construction, designed for lyophilization and freeze-drying.
Manufactured from Wheaton 33® low extractable borosilicate glass (meets ASTM Type I Class and USP Type I Standards).