Histomount, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17995-01 17995-05
101410-714EA 104.43 USD
101410-714 101410-716
Histomount, Electron Microscopy Science
Stains Microscopy and staining reagents
A mixture of xylene - mixed isomers and non hazardous ingredients to offer a very good synthetic mountant. Once dried the refractive index is 1.58.