Nuova™ Hot Plates, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
72358-10 72358-20 72358-22
102097-954EA 1038.32 USD
102097-954 102097-956 102097-958
Nuova™ Hot Plates, Electron Microscopy Sciences
Heaters Hot Plates
Hot plates with durable die cast aluminum case and corrosion resistant porcelain coated ceramic top.

  • Topside drip edge – protects internal components in case of spillage
  • Integral ring stand holder – accommodates 0.5" (13mm) support rod
  • Accommodates up to 15 lb loads
  • All unit included "PTFE" coated stir bar
  • Demand-Type thermostatic temperature control – excellent temperature stability ±5.0 °C (9 °F) @ 371 °C (700 °F)
  • Embedded heating elements – uniformity transfer heat to the top plate
  • Low temperature control: 38 °C (100 °F) – perfect choice for warming applications
  • All units max. operating temperature range is 540 °C

Applications:
General reagent heating
Evaporation – Sample drying
Heating TLC plates
Digestions

Recommended for use with glass vessels only.

Heating Surface: 10.25 x 10.25 in
Overall dimensions: 16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2 cm)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR