Iron(III) chloride 2% in aqueous solution for histology

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Ferric chlorideIron trichloride

15512-02
101410-656EA 66.13 USD
101410-656
Iron(III) chloride 2% in aqueous solution for histology
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
MDL Number: MFCD00011005
CAS Number: 7705-08-0
https://us.vwr.com/stibo/web/std.lang.all/48/22/28984822.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR