DiATOME® Ultra and Cryo Diamond Knife for AFM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
20-AFM-CDL 20-AFM-CDR 20-AFM-UDL 20-AFM-UDR 30-AFM-CDL 30-AFM-CDR 30-AFM-UDL 30-AFM-UDR
102680-568EA 6857.08 USD
102680-568 102680-570 102680-572 102680-574 102680-692 102680-694 102680-696 102680-698
DiATOME® Ultra and Cryo Diamond Knife for AFM, Electron Microscopy Sciences
Knives
These high quality Diatome diamond knives are not just perfect for producing ultrathin and semi-thin sections, but also for surfacing sectioning of all kinds of biological and industrial specimens.
  • Extremely smooth sample surfaces and best possible structure preservation
  • Available in 35° with a cutting range of 10-100 nm in 2 and 3 mm sizes
Instead of a section as for TEM, the specimen surface is investigated using AFM. In order to achieve the best results for AFM investigation, only the highest quality diamond knives should be used. Diatome ultra AFM and cryo AFM knives are specially tested to ensure that they meet the increased quality requirements of AFM investigation.