Applications include the study of microorganisms and tissue cell growth under strictly controlled conditions of temperature and agitation
Designed to provide the security of precise temperature control and uniformity for demanding incubation/refrigeration application without taking up a great deal of space. It uses Thermo-Electric (Peltier) refrigeration and heating with no compressor. Chamber air is gently and continuously recirculated at a rate that ensures the temperature consistency and uniformity of all test samples. Re-circulated air eliminates the possibility of contamination from outside air. Shatter resistant viewing window allows for inspection of samples without effecting the chamber environment.
Applications include the study of microorganisms and tissue cell growth under strictly controlled conditions of temperature and agitation. It performs membrane based assays/washing and hybridizations more efficiently. Microprocessor controls easily achieve desired rotating, rocking, stirring, shaking, temperature and time; all of which are displayed on large LED’s for accurate reproducible results. Timer displays elapsed time or remaining time until programmed off – up to 96 hours. Visible and audible alarms for both temperature and time. Equipped with RS232 port for connection to your computer.
It performs membrane based assays/washing and hybridizations more efficiently. Microprocessor controls easily achieve desired rotating, rocking, stirring, shaking, temperature and time; all of which are displayed on large LED’s for accurate reproducible results. Timer displays elapsed time or remaining time until programmed off – up to 96 hours. Visible and audible alarms for both temperature and time. Equipped with RS232 port for connection to your computer.