Ultra High Resolution Gold (<2 nm - 30 nm) on Carbon Test
Supplier: Electron Microscopy Sciences
79512-04 79512-03 79512-02 79512-01 79512-05
100494-962EA 903.51 USD
100494-962 100494-970 100494-968 100494-966 100494-964
Ultra High Resolution Gold (<2 nm - 30 nm) on Carbon Test
Microscopes
These gold on carbon calibration specimens have been specially produced to enable the ultra high-resolution capabilities of the current generation of high performance scanning electron microscopes to be assessed. In particular, the ultra small gold particle size (<1 nm - 20 nm) is designed for use with SEM 's equipped with field emission guns.
With gold particles ranging from 1 nm to 20 nm, this test is suitable for ultra high resolution SEM tests such as field emission electron source. A magnification of at least 80,000x is required for this test.