Silver Adhesives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12641 12640
100503-772EA 15.81 USD
100503-772 100503-770
Silver Adhesives, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Silver adhesive in liquid and paste formats.
  • Silver liquid is fast drying
  • Silver paste is applied with micro spatula or toothpick
  • Silver extender available
Liquid; Colloidal Silver: 1-Methoxy-2-propanol base
Paste; Colloidal Silver: Clear Lacquer base. Thick base – Ideal for non-flowing required. Easily applied with micro spatula or wooden toothpick. Particle size ranging 0.4 – 1 µm, 80% are less than 1 µm. Cure in 16 to 20 hours at room temperature or 30 minutes at 125 – 150 °C
Fast drying. Average grain size less than 1 µm and Silver content is 60%. Sheet resistance is 0.02 ohm per square inch at 1 mil thickness. Comes with a brush attached to the cap. Service temperature is 30 minutes at 200°C.