Stage Graticules and Micrometers for Transmitted Light, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68039-02 68043-20 68039-11 68042-08 68039-05 68044-12 68039-16 68039-22 68039-04 68040-01 68041-48 68043-21
100499-300EA 455.16 USD
100499-300 100499-318 100499-306 100499-316 100499-304 100499-324 100499-308 100499-310 100499-302 100499-312 100499-314 100499-320
Stage Graticules and Micrometers for Transmitted Light, Electron Microscopy Sciences
Graticules, Reticles and Grids
The stage graticules are for the routine calibration of eyepiece patterns when alternating between objectives or using the graticule in different microscopes.
  • Robust construction
  • Ideal for student use and for instructional purposes
The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75×24×2 mm thick.