Centrifugal Filter Devices - Costar Brand , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67030-62 67031-69 67030-60 67030-61 67031-70 67030-63
100500-514CS 561.46 USD
100500-514 100501-760 100500-510 100500-512 100501-762 100500-516
Centrifugal Filter Devices - Costar Brand , Electron Microscopy Sciences
Filters Centrifugal Filters
Spin-X centrifuge tube filters utilize centrifugation for the filtering of small volumes.
  • Filter samples in microcentrifuges
  • Choice of cellulose or nylon membrane
  • For volume up to 500 µl
The unit consists of a polypropylene filter insert and a centrifuge tube. They have a cellulose acetate membrane for aqueous solutions which is ideal for the removal of bacteria, particles, and cells from media. They are also effective for DNA removal from agarose or acrylamide gels. Spin-X LC units have a nylon membrane for applications such as HPLC sample preparation. Both have retailed volume of less than 5 µl, and withstand relative centrifugal force (RFC) to 16000×g.