Wooden Sticks; Tongue Depressors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72322 72320
100500-652BX 37.41 USD
100500-652 100488-474
Wooden Sticks; Tongue Depressors, Electron Microscopy Sciences
Tongue Depressors
These sticks are made from close-grained northern white birch because of its ability to provide a smooth and polished surface.
Used for mixing, stirring, or scraping epoxy resins.