JB-4 Plus™ Embedding Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
JB-4 Plus offers all of the same features as JB-4 except it produces less of an exothermic reaction than JB4 which is good for temperature sensitive tissues
JB-4 Plus Kit Consists of 500 mL Solution A , 15 mL Solution B , and 8 g Benzoyl Peroxide Catalyst.
It produces harder blocks which is ideal for dense samples, such as bone.