Graphite Adhesives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12661 12660 12650
100503-778EA 14.45 USD
100503-778 100503-776 100503-774
Graphite Adhesives, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Water based and isopropal based graphite adhesives.
  • Water based
  • Isopropal based
  • Graphite extender available
Water based has a flat surface texture. The average flake size is 1 micron. Service temperature is 306°C. Isopropal based has a flat surface texture. The average flake size is 1 micron. Service temperature is 204°C.