Anti-IgG Antibody

Supplier: Electron Microscopy Sciences
25801 25806 25808 25800 25803 25807 25809 25802 25804
100488-014EA 430.9 USD
100488-014 100488-024 100488-028 100488-012 100488-022 100488-018 100488-026 100488-030 100488-016 100488-020
Anti-IgG Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR