Cytoseal Mounting Medium, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
18006 18007 18008
102090-994EA 60.6 USD
102090-994 102090-996 102090-998
Cytoseal Mounting Medium, Electron Microscopy Sciences
Stains Microscopy and staining reagents
CytoSeal comes in a Controlled Drop Dispenser, with varying viscosities
Premium quality formulated from the highest quality advanced acrylic resin. It will not become brittle and crack, nor will it discolor or yellow with age. CytoSeal dries rapidly, allowing examination soon after application. It can be used with oil immersion objectives and in fluorescent procedures. Slides will not stick together and the medium will not "cold flow" to the edge of the slide during long term storage. CytoSeal can be dissolved in toluene or xylene. An antioxidant has been added to the formulation to prevent fading of slides.
CytoSeal comes in a Controlled Drop Dispenser, with varying viscosities. CytoSeal 60, which has a viscosity of 60cps, is intended primarily for use with cover glass. CytoSeal 280, which has a viscosity of 280cps is intended for use where minimal spread of medium is required.
CytoSeal 60, which has a viscosity of 60cps, is intended primarily for use with cover glass. CytoSeal 280, which has a viscosity of 280cps is intended for use where minimal spread of medium is required.