Safeknit™ Cut-Resistant Gloves, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71065-06 71065-08 71065-09 71065-10 71065-11 71066-06 71066-07 71066-08 71066-09 71066-10 71066-11
102097-284EA 38.07 USD
102097-284 102097-288 102097-290 102097-292 102097-294 102097-296 102097-298 102097-300 102097-302 102097-304 102097-306
Safeknit™ Cut-Resistant Gloves, Electron Microscopy Sciences
Gloves
Safeknit™ is a blend of abrasion-resistant Spectra® fiber and other synthetics
  • Extreme comfort and productivity
  • These gloves are neither puncture proof nor water resistant
The mix of these fibers form a rugged cut resistant barrier that provides the histologist sensitivity for manipulation. They should not be used to protect against intentional cutting. Avoid use near moving blades. For applications which are related to fluids where a water resistant glove is required, we recommend either double gloving or using nitrile gloves.
Available in twoMore Product Information