Dumont Tweezers Style 12, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0305-12NI-CO 0305-12NI-PO
102091-732EA 51.29 USD
102091-732 102091-734
Dumont Tweezers Style 12, Electron Microscopy Sciences
Forceps
Thin, 120 mm tips. Ideal to manipulate delicate pieces.
Nickel is a 100% antimagnetic alloy composed of Cu, Ni, Mn and Zn. lt is widely appreciated as it does not damage the components handled by it.