Dumont Tweezers Style 2, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72874-D 72874-DZ
100493-846EA 82.27 USD
100493-846 100503-108
Dumont Tweezers Style 2, Electron Microscopy Sciences
Forceps
Strong tips.
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400°C (DIN 50914) and is suitable for autoclave sterilization at 180°C.