Glover Atraumatic Bulldog Clamps, Roboz Surgical
Supplier: Roboz Surgical
RS-7530 RS-7541
100188-900EA 386.31 USD
100188-900 100188-868
Glover Atraumatic Bulldog Clamps, Roboz Surgical
Clamps General Purpose Clamps
The Bulldog clamp is a useful instrument for stopping blood flow to a particular organ of interest
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled. Tension Adjusting Screw.