Anti-AdV Mouse Monoclonal Antibody [clone: M58 + M73]

Supplier: Diagnostic BioSystems

MOB355 MOB355-05
95024-222EA 424.79 USD
95024-222 99990-664
Anti-AdV Mouse Monoclonal Antibody [clone: M58 + M73]
Antibodies
This antibody reacts with adenovirus. The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46 kDa) with multifunctional capabilities and forms a specific complex with the retinoblastoma tumor suppressor gene product. The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region.

Type:
Antigen: AdV
Clonality: Monoclonal
Clone: M58 + M73
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR