Clarity Lancets and Lancing Device, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-GL6 DTG-GL7
95064-157CS 1726.17 USD
95064-157 75871-632
Clarity Lancets and Lancing Device, Clarity Diagnostics
Blood Lancets
Clarity lancing device to be used with Clarity regular twist top 30G lancets.
  • 1-5 skin depth settings
  • Limited 2 year warranty on lancing device
  • For single patient use
  • Lancet ejector
  • Ergonomic design
30G sterile one use twist top lancets are virtually painless.