HYGEN™ Microfiber Dry Room Mop
Supplier: Rubbermaid
HYGEN™
RCPQ412GRE RCPQ412GRE
19758-829EA 33.58 USD
19758-829 19758-828
HYGEN™ Microfiber Dry Room Mop
Mops
High-pile, 100% microfiber collects and holds dust and dirt mechanically and electrostatically for superior dusting results.
Dense 3 to 5 µm quality fiber removes more dust, dirt and microbes than conventional cleaning products. Advanced hook-and-loop backing; holds-up under harsh laundering conditions and attaches pads more securely. Double finished edges retain mop shape with use over time. Color coded products help reduce cross contamination by differentiating area of use.