HYGEN™ High-Performance Damp Mops, Rubbermaid®
Supplier: Janitorial Supplies
RCPQ410RED RCPQ411BLU
19758-825EA 55.88 USD
19758-825 19758-827
HYGEN™ High-Performance Damp Mops, Rubbermaid®
Mops
Premium split nylon/polyester blend microfiber provides optimal damp mopping performance
Dense 3 - 5 micrometer quality fiber removes more dust, dirt and microbes than conventional cleaning products. Advanced hook-and-loop backing; holds-up under harsh laundering conditions and attaches pads more securely. Double finished edges retain mop shape with use over time. Color coded products help reduce cross contamination by differentiating area of use. Patent Pending Zig-Zag design packs 17% more split microfiber into each mop. Durable mop construction; third party tested tough. Withstands more than 500 launderings, 200 with bleach. Proven to clean floors 45% better than string mops, 25% better than conventional microfiber.